Nalgene® Oak Ridge Centrifuge Tubes, Polycarbonate, Thermo Scientific
Supplier: Thermo Fisher Scientific
3118-0010 3118-0030 3118-0028 3118-0050 3118-0085 3138-0016 3138-0030 3138-0050
21009-310CS 2162.62 CAD
21009-310 21009-320 21009-331 21009-342 21009-353 21010-343 21010-385 21010-400
Nalgene® Oak Ridge Centrifuge Tubes, Polycarbonate, Thermo Scientific
Tubes Centrifuge Tubes
Transparent autoclavable polycarbonate tubes.
  • Polypropylene screw caps
  • Leakproof closures provide safe containment
  • Withstand up to 50000 ×g in standard and refrigerated centrifuges
Place cap loosely on tube when autoclaving, without engaging threads.
Note: For proper performance, fill tubes to at least 80% of the total capacity.
Certifications: Meet OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens.
Ordering information: Tubes with sealing caps include a silicone gasket in addition to the tube and screwMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO14001 (Certificate Registration No. EMS 752603)