Ethanol denatured 90% (denatured with methanol and isopropanol)

Supplier: G-Biosciences

Synonyms: Ethyl alcohol denatured

786-2077
77540-212EA 40.87 CAD
77540-212
Ethanol denatured 90% (denatured with methanol and isopropanol)
Ethanol denatured

Anhydrous Ethanol (denatured) is used as a mobile phase in High Performance Liquid Chromatography (HPLC) and Liquid Chromatography coupled with Mass Spectrometry (LCMS).


  • An alcohol like order


Miscible with water and most organic solvents

Formula: H₃CCH₂OH
MW: 46.07 g/mol
MDL Number: MFCD00003568
CAS Number: 64-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR