Nalgene® Oak Ridge Centrifuge Tubes, Teflon® FEP, Thermo Scientific
Supplier: Thermo Fisher Scientific
3114-0010 3114-0030 3114-0050
21009-422CS 926.75 CAD
21009-422 21009-455 21009-477
Nalgene® Oak Ridge Centrifuge Tubes, Teflon® FEP, Thermo Scientific
Tubes Centrifuge Tubes
For high-speed centrifugation of aggressive chemicals.
  • Autoclavable and leakproof
  • Resists any acid, base, or solvent
  • Withstand temperatures from –100 to 150 °C (–148 to 302 °F)
  • Withstand upto 50000 ×g in refrigerated centrifuges
Excellent for lipid and phenol extractions, as well as biomolecular applications using nucleic acids, proteins, and more. Provide storage for expensive or aggressive samples. Before autoclaving, place closure on container without engaging threads.
Note: FEP centrifuge ware must be completely filled and refrigerated forMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable