Drying Tube, [ST] 14/20 Joint, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences
970440
80093-774CS 81.08 CAD
80093-774
Drying Tube, [ST] 14/20 Joint, Wheaton, DWK Life Sciences
Tubes Drying Tubes
Constructed of Wheaton 33® low extractable borosilicate glass.
The design of the tube eliminates the ball and socket connecting the scrubber and absorber components. The apparatus follows the silver diethyldithiocarbamate method described in the 18th edition of Standard Methods for the Examination of Water and Wastewater.
Certifications: Tube meets ASTM Type I Class A and USP Type I standards.