U-Shaped Drying Tubes, Chemglass
Supplier: Chemglass
CG-1296-01 CG-1296-14
80067-994EA 206.12 CAD
80067-994 80067-998
U-Shaped Drying Tubes, Chemglass
Tubes Drying Tubes
Tubes feature a [ST] lower inner joint and include a rubber stopper and glass inlet tube.