Decolorizer (3% Acid Alcohol)
Supplier: G-Biosciences
786-2113
786-2114
77540-284KT
209.93
CAD
77540-284
77540-286
Decolorizer (3% Acid Alcohol)
Stains
Microscopy and staining reagents
Decolorizer is used with the Kinyoun or Ziehl-Neelson Carbolfuchsin staning procedure.
- An aqueous solution of hydrochloric acid and ethanol
It differentiates acid-fast bacteria from non-acid-fast bacteria