Auramine-Rhodamine T solution
Supplier: G-Biosciences
786-2109
786-2110
77540-276EA
78.61
CAD
77540-276
77540-278
Auramine-Rhodamine T solution
Auramine-Rhodamine T
Auramine-Rhodamine T is used in the Fluorescent microscopic system.
- Quick staining dye
It is used for the detection of mycobacteria