Auramine O solution

Supplier: G-Biosciences

786-2106 786-2107
77540-270EA 48.74 CAD
77540-270 77540-272
Auramine O solution
Auramine O

Auramine O is used in the Fluorescent microscopic system.


  • Quick staining kit


It is used for the detection of mycobacteria

Formula: C₁₇H₂₂ClN₃
MDL Number: MFCD00012484
CAS Number: 2465-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR