Gram′s safranin solution
Supplier: G-Biosciences
Synonyms:
Safranin T solution
786-2099
786-2100
786-2101
77540-256EA
402.46
CAD
77540-256
77540-260
77540-258
Gram′s safranin solution
Gram′s safranin solution
Gram Safranin acts as a counterstain .
- An aqueous dye solution
It is used in Gram staining to differentiate gram-positive from gram-negative bacteria
Formula:
C₂₀H₁₉ClN₄ MW: 350.85 g/mol |
MDL Number:
MFCD00011759 CAS Number: 477-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...