InLab® Redox Combination Electrodes, METTLER TOLEDO®
Supplier: Mettler Toledo
51343200 51343202
CA97001-684EA 948.89 CAD
CA97001-684 CA97001-686
InLab® Redox Combination Electrodes, METTLER TOLEDO®
Electrodes Redox Electrodes
Electrodes measure oxidation/reduction power of a solution
  • Platinum ring indicator
  • Safelock™ closing ring
  • ARGENTHAL™ lead-off and silver ion trap
  • S7 screw head connector
They feature a platinum ring indicator, a Safelock™ closing ring for easy refilling and complete sealing, and an ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures. S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters.