Corning® Thermowell® GOLD and Thermowell PCR Tubes, Corning
Supplier: Corning
6530 6531 3744 3745 3746 3747 3749 6571 3750
29442-340CS 152.48 CAD
29442-340 29443-340 33502-114 33502-116 33502-118 33502-120 33502-122 53495-756 33502-124
Corning® Thermowell® GOLD and Thermowell PCR Tubes, Corning
Tubes PCR Tubes
Manufactured from ultra-pure, medical grade PP suitable for RT-PCR.
  • Available with flat caps for easy piercing and marking, or domed caps for efficient heat transfer
  • Ultra-thin walls of uniform thickness accelerate heat transfer for shorter cycle times and higher product yields
  • 100% tested for well integrity
  • Certified DNase- and RNase-free and endotoxin-safe on a lot-by-lot basis
  • Autoclavable at 121 °C
  • Able to withstand centrifugation to 10000×g
Tubes are designed with longer cap flanges and sealed with a full-length insertion cap to safeguard against evaporation and to prevent accidental cap opening during thermal cycling.
Ordering information: Assorted pack includes the following colors: blue, green, orange, red, violet, and yellow.