VWR® PCR Tubes and Caps
Supplier: VWR International
20170-010 20170-012 76318-804 76446-868 76446-876 76446-878 76446-880
20170-010CS 1421.44 CAD
20170-010 20170-012 76318-804 76446-868 76446-876 76446-878 76446-880
VWR® PCR Tubes and Caps
Tubes PCR Tubes
Ideal for all PCR protocols, these 0.2 ml tubes and caps are made from polypropylene and are compatible with most popular thermal cycler blocks.
  • Autoclavable
  • Chemically inert
They feature smooth, thin, uniform walls for accurate thermal transfer. Certified free of RNase, DNase, and human DNA (lot-by-lot certificates available). Tubes are available with bubble-top or flat-top caps, both of which fit precisely to ensure a positive, contamination-free environment.