VWR® PCR 12-Well Tube Strips and Caps
Supplier: VWR International
53509-300 53509-302 53509-306 76446-866
53509-300CS 2713.07 CAD
53509-300 53509-302 53509-306 76446-866
VWR® PCR 12-Well Tube Strips and Caps
Tubes PCR Tubes
Ideal for all PCR protocols, these 0.2 ml tubes and caps are made from polypropylene and are compatible with most popular thermal cycler blocks.
  • Chemically inert
  • Autoclavable
  • Caps ensure a contamination-free environment
They feature smooth, thin, uniform walls for accurate thermal transfer. Certified free of RNase, DNase, and human DNA (lot-by-lot certificates available). Caps fit precisely.
Ordering information: Tube strips with separate cap strips (53509-306) are packed with 80 tubes and 80 caps in separate bags inside a box.