Pressure Equalizing Funnels, Graduated with PTFE Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
9493-40 9493-06 9493-14 7297-37 9493-05 7297-39 9493-08 9493-10 9493-15 9493-07 7297-33 9493-20 7297-45 7297-31 7297-35 9493-03
89074-972EA 658.42 CAD
89074-972 89058-738 89074-982 89074-970 89074-980 89074-976 89074-986 89085-122 89074-974 89074-984 89074-978 89058-740 89058-742 89058-744 89058-736 89058-746
Pressure Equalizing Funnels, Graduated with PTFE Stopcock, Ace Glass Incorporated
Funnels Separating Funnels
Cylindrical with standard taper joint top and bottom, drip stem, pressure equalizing arm and double scale graduations, calibrated from stopcock
Drip stem does not extend beyond joint, thereby eliminating possible source of breakage.
Ordering information: Stoppers are not supplied. For stoppers or replacement stopcock plugs, contact your Avantor representative.