Separatory Funnels, Squibb, 1:5 PTFE Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
7228-77 7228-79 7228-66 7228-68 7228-64 7228-70 7228-72 7228-62 7228-60 7228-76
89058-554EA 1173.71 CAD
89058-554 89058-556 89058-680 89058-682 89058-710 89058-790 89058-792 89074-950 89075-014 89058-552
Separatory Funnels, Squibb, 1:5 PTFE Stopcock, Ace Glass Incorporated
Funnels Separating Funnels
Squibb-shaped separatory funnel with 1:5 PTFE stopcock on the bottom outlet and [ST] PTFE stopper in neck
Manufactured from borosilicate glass.
Ordering information: For replacement stoppers or stopcock plugs, contact your Avantor representative.
Order Now
SPECIFICATIONS
Environmentally Preferable