Mµlti™ µltraStrips, Sorenson BioScience
Supplier: Sorenson Bioscience
16330 16340 16360 16280 16290 16300 16310
53550-116CS 642.67 CAD
53550-116 53550-118 53550-122 53550-126 53550-128 53550-130 53550-132
Mµlti™ µltraStrips, Sorenson BioScience
Tubes PCR Tubes
Ideal for PCR protocols, thin-walled µltraStrips feature dual connectors between the reaction tubes and caps for added sample security.
  • certified RNase-, DNase-free
  • Available in several colors for easy identification
  • Autoclavable
The 8×0.2 ml format fits precisely in thermal cycler blocks for uniform heat transfer. µltraStrips are packed with matching cap strips for added convenience. µltraStrips can be easily cut and used as individual tubes.