VWR® PCR Strip Tubes & Caps 8-Strip
Supplier: VWR International
20170-000 20170-002 20170-004 53509-304 82006-606 83009-684 89096-722 93001-118 93001-120 89401-570 76318-796 76318-798 76318-800 76318-802 76402-850 76402-852 76402-854 76402-856 76446-858 76446-860 76446-862 76446-864 76446-870 76446-872 76446-874
20170-000CS 955.21 CAD
20170-000 20170-002 20170-004 53509-304 82006-606 83009-684 89096-722 93001-118 93001-120 89401-570 76318-796 76318-798 76318-800 76318-802 76402-850 76402-852 76402-854 76402-856 76446-858 76446-860 76446-862 76446-864 76446-870 76446-872 76446-874
VWR® PCR Strip Tubes & Caps 8-Strip
Tubes PCR Tubes
Ideal for all PCR protocols, these 0.1 and 0.2 ml tubes and caps are made from polypropylene and are compatible with most popular thermal cycler blocks.
  • Tubes and caps are chemically inert and autoclavable
  • Smooth, thin, uniform walls for accurate thermal transfer
  • Caps fit precisely to ensure a positive, contamination-free environment
Tube strips are certified free of RNase, DNase, and human DNA (lot-by-lot certificates available).
Ordering information: Tube strips with separate bubble cap strips are packed with 125 strips of 8 tubes and 125 strips of 8 caps in separate bags inside a box. Tube strips for real time PCR come with individually attached, optically clear caps.