SICURIX® Ribbon Cartridge
Supplier: Essendant
Fargo®
SRX45000
77531-646EA 240.97 CAD
77531-646
SICURIX® Ribbon Cartridge
Printer Ribbons
The SICURIX® ribbon cartridge is an alternative for Fargo (45000). Full-color ribbon cartridge is especially designed for printing a combination of full-color images and one-color text or bar codes in the Fargo DTC1000 Card Printer/Encoder. The Fargo 45000 color ribbon features a resin black and clear overlay panel and includes a built-in cleaning roller. YMCKO ribbon yields 250 images.
  • Designed for use with the Fargo DTC1000 Card Printer/Encoder
  • Features a resin black and clear overlay panel
  • Includes a built-in cleaning roller
Print technology: Dye sublimation and thermal transfer