KIMBLE® Pressure Caps for NMR Sample Tubes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
897093-0001 897095-0012 897095-0021 897095-0061 897095-0071 897095-0081 897095-1011 897095-1021 897095-0001 897095-0011 897095-1001
KT897093-0001PK 34.71 CAD
KT897093-0001 KT897095-0012 KT897095-0021 KT897095-0061 KT897095-0071 KT897095-0081 KT897095-1011 KT897095-1021 KT897096-0003 KT897095-0001 KT897095-0011 KT897095-1001
KIMBLE® Pressure Caps for NMR Sample Tubes, DWK Life Sciences
Tubes NMR Tubes
Tight-fitting caps withstand vaporization pressure buildup within the sample tube.
Caps are precision machined and available in varied polyethylene sizes.
Ordering information: Pack of assorted 10 mm caps (KT897095-1011) contains 25 each of red, black, green, and yellow.