Rotary Evaporator Traps, Anti-Climb, Ace Glass Incorporated
Supplier: Ace Glass
6706-05 6706-13 6706-17 6706-21 6706-09
89055-884EA 405.42 CAD
89055-884 89055-888 89055-890 89055-892 89055-886
Rotary Evaporator Traps, Anti-Climb, Ace Glass Incorporated
Vacuum Traps
Upper tubulation prevents migration of film and foam directly into the condenser
Manufactured from borosilicate glass.