KONTES® Rotary Evaporator Traps, with Drain Holes, Kimble Chase
Supplier: Kimble Chase
KONTES®
570205-0124 570205-2514 570205-0114
KT570205-0114CS 506.26 CAD
KT570205-0114 KT570205-0124 KT570205-2514
KONTES® Rotary Evaporator Traps, with Drain Holes, Kimble Chase
Vacuum Traps
Designed for use where foaming is likely to occur and be drawn into the condenser
Traps have an inner vapor tube at the bottom that features two 4 mm (3/16") drain holes at its base. Drain holes are positioned 180° to each other and 90° to the upper holes for faster drainage of trap contents back into the evaporation flask. Each flask has a 24/40 top outer [ST] ground joint.