Rotary Evaporator Bump Traps, Chemglass
Supplier: Chemglass
CG-1321-03 CG-1321-102 CG-1321-252 CG-1320-01 CG-1320-03 CG-1320-04 CG-1320-06 CG-1320-09 CG-1321-01 CG-1321-04 CG-1321-06 CG-1321-09 CG-1321-11 CG-1321-12 CG-1321-14 CG-1322-01 CG-1322-03 CG-1322-04 CG-1322-06 CG-1322-09
80068-082EA 467.54 CAD
80068-082 89168-682 89168-684 80068-058 80068-060 80068-062 80068-064 80068-068 80068-080 80068-084 80068-086 80068-090 80068-094 80068-096 80068-100 80068-102 80068-104 80068-106 80068-108 80068-112 10170-350
Rotary Evaporator Bump Traps, Chemglass
Vacuum Traps
Rotary Evaporator Bump Traps is used between the vapor tube and evaporation flask on a rotary evaporator to prevent contents of the flask from being drawn into the condenser in the event of bumping.
Modified bump traps provide expanded volume for reactions that have a tendency to foam excessively. The inner vapor tube is sealed to the top of the trap to prevent entry into the condenser. Traps with drain holes feature two 4mm holes at the base of the inner tube to allow solvent to drain back into the evaporation flask.