KONTES® Soxhlet Extraction Apparatus, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
586000-0023 586000-0024 586000-0025 586000-0021 586000-0022
KT586000-0023EA 851.92 CAD
KT586000-0023 KT586000-0024 KT586000-0025 KT586000-0021 KT586000-0022
KONTES® Soxhlet Extraction Apparatus, Kimble Chase, DWK Life Sciences
Extractors Soxhlet and Soxhlet-Type Extractors
Used for the continuous extraction of solids with a suitable solvent
Soxhlet-type apparatus with Allihn Condenser features a bulb-type Allihn condenser, protected siphon tube, enlarged vapor tube, and sloping seals to prevent the trapping of liquid. The extractor is connected to the condenser and to the flask by [ST] joints. Suitable for use with paper, alundum, or fritted glass extraction thimbles (not included). Extractor Only components are also available.
Order Now
SPECIFICATIONS
Environmentally Preferable