KIMAX® Extraction Apparatus with Allihn Condenser, Flask, and Soxhlet Extraction Tube, [ST] Joints, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
24005-30 24005-40 24005-50 24027-30 24027-40 24071-50 24027-50 24071-30 24071-40
89001-768CS 1246.42 CAD
89001-768 89001-770 89001-772 89001-774 89001-776 89001-784 89001-778 89001-780 89001-782
KIMAX® Extraction Apparatus with Allihn Condenser, Flask, and Soxhlet Extraction Tube, [ST] Joints, Kimble Chase
Extractors Soxhlet and Soxhlet-Type Extractors
Extraction apparatus consists of an Allihn condenser, flask and Soxhlet extraction tube
All are equipped with interchangeable [ST] ground joints.
Order Now
SPECIFICATIONS
Environmentally Preferable