Neptune® Microcentrifuge Tubes with Attached Flat Caps, Biotix
Supplier: Biotix
63300631 63300632 63300637 63300638 63300641 63300642 63300643 63300645 63300646
89126-710CS 1057.27 CAD
89126-710 89126-714 89126-716 89126-718 89126-720 89126-722 89126-724 89126-726 89126-728 89126-730 89134-500 89134-502 89134-504 89134-506
Neptune® Microcentrifuge Tubes with Attached Flat Caps, Biotix
Tubes Microcentrifuge Tubes
Tubes are designed for general laboratory use and storage of aqueous solutions.
Exceptional optical clarity and well distinguished graduation marks allow easy visual reference of sample. Will withstand autoclaving at 121 °C (250 °F) at 15 psi for 15 min. Rated to RCF of 20000×g for 15 min. Tubes are frosted on the top and sides for convenient sample labeling.
Certifications: CE certified. IVD labeled. Certified free of DNA, DNase, RNase, and endotoxins (pyrogens). Manufactured under stringent controls in an ISO 9001 certified facility. Every lot is tested for precision assuring maximumMore Product Information