Microcentrifuge Tubes, Polypropylene, Greiner Bio-One
Supplier: Greiner Bio-One
616201 623201 667201 717201 717261 742270 616261 722261 616283 716201
82050-504CS 199.83 CAD
82050-504 82050-514 82050-530 82050-934 82051-120 82051-122 82051-124 82051-160 89204-782 89217-356 10803-072 10026-350
Microcentrifuge Tubes, Polypropylene, Greiner Bio-One
Tubes Microcentrifuge Tubes
Microcentrifuge tubes are manufactured from highly transparent, inert polypropylene resin that offers superior chemical resistance and thermal stability.
  • Polypropylene construction
  • Highly transparent and inert
  • Superior chemical and temperature resistance
  • Autoclavable
  • Available nonsterile or sterile
Color-coded tubes are available for easy and quick identification.
Tubes are designed to suit most analyzer systems, including Eppendorf®, Viatron, Roche®, and Beckman® systems.
All 1.5 ml tubes feature molded graduations. Ideal solution for secure,More Product Information