VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
87003-290 87003-292 87003-294 87003-296 87003-298 87003-300 87004-254 87004-256 87004-258 87004-260 87004-262 87004-264 87004-266 87004-268 87004-270 87004-272 87004-274 87004-276 87004-278 87004-280 87004-282
87003-290CS 1706.5 CAD
87003-290 87003-292 87003-294 87003-296 87003-298 87003-300 87004-254 87004-256 87004-258 87004-260 87004-262 87004-264 87004-266 87004-268 87004-270 87004-272 87004-274 87004-276 87004-278 87004-280 87004-282
VWR® Microcentrifuge Tubes, PP
Tubes Microcentrifuge Tubes
Polypropylene tubes feature superior clarity and a positive seal design that withstands repeated cap closures and will not pop open during boiling.
  • Secure positive seal design
  • Tubes fit all standard rotors
  • Certified RNase-, DNase-, and pyrogen-free
  • Certified PCR inhibitor- and human DNA-free
  • Autoclavable
Temperature range is from –80 to 121 °C (–112 to 250 °F).
Ordering information: Rainbow packs consist of red, blue, green, yellow, and purple tubes.
Order Now
SPECIFICATIONS
Environmentally Preferable