BRAND Microcentrifuge Tubes and Racks, BrandTech
Supplier: Brandtech
780507 780550 780540 780536 780546
80087-078PK 415.27 CAD
80087-078 80087-080 80087-084 80087-095 80087-096 80087-091 80087-087 80087-089 10215-490 10755-230 10755-232
BRAND Microcentrifuge Tubes and Racks, BrandTech
Tubes Microcentrifuge Tubes
Manufactured from virgin polypropylene, these highly translucent microcentrifuge tubes are free from flash, air bubbles, and streaks.
  • 0.5 ml, 1.5 ml and 2.0 ml sizes available
  • Different colors available
  • Lid closure models withstand up to 30000 RCF
They are produced with uniform wall thickness to consistently withstand centrifugal forces. Attached caps are flat with consistent thickness for trouble-free piercing and form a tight, leak-free seal that is easy to open, even when wearing gloves. Tubes fit most common rotors and may be spun up to 30000 RCF (More Product Information