Axygen® Microcentrifuge Tubes without Caps, Graduated, Corning
Supplier: Corning
MCT-150-NC MCT-200-NC
10011-706CS 841.99 CAD
10011-706 10011-748
Axygen® Microcentrifuge Tubes without Caps, Graduated, Corning
Tubes Microcentrifuge Tubes
Clear polypropylene capless microcentrifuge tubes have ultrasmooth bottoms to allow for 100% visibility and no sharp 'gates' to cause cuts and scratches.
  • Certified RNase-, DNase-, and pyrogen-free
Tubes come with frosted side surface areas for writing and extremely accurate and visible graduation markings.
Ordering information: Tubes are supplied without caps.