Axygen® Screw Top Microcentrifuge Tubes without Caps, Polypropylene, Corning
Supplier: Corning
ST-050 ST-050-SS ST-050-SS-X ST-050-X ST-150 ST-150-SS ST-150-SS-X ST-200 ST-200-SS ST-200-SS-X ST-200-X ST-200-C-S ST-150-C-S ST-45-SS
10011-652CS 160.16 CAD
10011-652 10011-654 10011-656 10011-658 10011-660 10011-662 10011-664 10011-666 10011-668 10011-670 10011-672 10011-674 47751-112 47751-114 89179-786
Axygen® Screw Top Microcentrifuge Tubes without Caps, Polypropylene, Corning
Tubes Microcentrifuge Tubes
Polypropylene screw cap microcentrifuge tubes feature an alignment tab to align the tube with the centrifuge rotor for multiple applications, such as desalting.
This feature allows the tube to be re-spun without disturbing the partially pelletized sediment.
Certifications: Certified RNase-, DNase-, and pyrogen-free.
Ordering information: For compatible screw caps, see the 10011-394 series.