Axygen® Screw Top Microcentrifuge Tubes with Caps, Polypropylene, Corning
Supplier: Corning
SCT-050-C-S SCT-150-A-S SCT-150-C SCT-150-C-S SCT-200-A-S SCT-200-C SCT-200-C-S
10011-488CS 3044.15 CAD
10011-488 10011-496 10011-536 10011-542 10011-544 10011-592 10011-598 10011-600 10011-642
Axygen® Screw Top Microcentrifuge Tubes with Caps, Polypropylene, Corning
Tubes Microcentrifuge Tubes
These conical-bottom screw cap microcentrifuge tubes serve a broad range of laboratory needs including collection, pressing, centrifugation, long-term storage, and OEM packaging and shipping of restriction enzymes or other DNA/RNA modifying reagents.
Color-coded caps provide a syringe port that is easily pierceable for radioisotope labeling and fraction collecting. The threaded O-ring design ensures a uniform closure allowing centrifugation up to 20000×g. Microcentrifuge tubes have ribs on the cap and tube to fit the Axyrack™ Microtube Racks to allow single-handed opening and closing. Polypropylene tubes are certified RNase-, DNase-, and pyrogen-free.
Ordering information: Tubes without ribs are also available; contact your sales representative for information.
All microtubes include non-looped caps with O-rings. Presterilized microtubes are supplied with cap attached.