Orion™ Triode™ 3-in-1 pH/Automatic Temperature Compensation Probe, Thermo Scientific
Supplier: Thermo Fisher Scientific
9107BNMD 9107WMMD 9107BN
10010-732EA 1416.12 CAD
10010-732 10010-788 21000-042 23198-210 52458-202
Orion™ Triode™ 3-in-1 pH/Automatic Temperature Compensation Probe, Thermo Scientific
Electrodes pH Electrodes
Operate the highly-versatile Orion™ Triode™ 3-in-1 pH/Automatic Temperature Compensation Probe in either the field or a plant.
  • pH range of 0 to 14
  • Wick junction
  • Ag/AgCl sealed internal reference system
Device contains a low-maintenance, gel-filled electrode with built-in ATC.