Orion™ Dissolved Oxygen/BOD Probe, Thermo Scientific
Supplier: Thermo Fisher Scientific
970801 970802 970803
34111-272EA 165.54 CAD
34111-272 34111-294 34111-308 CA10010-828
Orion™ Dissolved Oxygen/BOD Probe, Thermo Scientific
Electrodes Dissolved Oxygen Electrodes
Make any pH meter a dissolved oxygen meter with the Orion™ Dissolved Oxygen/BOD Probe.
  • Polarographic O₂ sensor plus solid-state circuitry in probe top; converts current to voltage for direct DO readout
  • Plug probe into benchtop or portable pH meter, read DO over 0–14 ppm range
  • Accuracy at 15 to 35 °C (59 to 95 °F): ±0.05 ppm/2% of reading, whichever is greater
  • Dual thermocompensator corrects for membrane permeability and O₂ solubility
  • Ideal for BOD testing; overflow funnel supplied fits 300 ml BOD bottle; built-in magnetic stir bar
  • Screw-on membrane/
More Product Information