VWR® Microcentrifuge Tubes with Socket Screw-Cap
Supplier: VWR International
89004-280 89004-282 89004-284 89004-286 89004-288 89004-290 89004-292 89004-294 89004-296 89004-298 89004-300 89004-302 89004-304 89004-308 89004-312 89004-316 89004-320 89004-324
89004-280CS 3283.19 CAD
89004-280 89004-282 89004-284 89004-286 89004-288 89004-290 89004-292 89004-294 89004-296 89004-298 89004-300 89004-302 89004-304 89004-308 89004-312 89004-316 89004-320 89004-324
VWR® Microcentrifuge Tubes with Socket Screw-Cap
Tubes Microcentrifuge Tubes
These clear high grade gamma resistant PP tubes are ideal for sample processing applications.
  • Autoclavable at 121 ℃ and freezable to –80 ℃, and excellent for frozen storage
  • Caps accept color coded inserts, and easily write on the frosted side of insert
  • O-rings are made from silicone to insure a leakproof seal
  • RNase-, DNase-, and endotoxin-free
  • Sterile tubes are sterilized by irradiation
Excellent for freezer storage or secure sample handling. Screw caps incorporate silicone O-rings for a leakproof seal and accept color-coded inserts. Inserts snap intoMore Product Information