VWR® Sample Library Tubes and Closures, 1.2 ml
Supplier: VWR International
82006-698 82006-700 82006-702 93000-040 93000-042 93000-044
82006-698CS 3016.34 CAD
82006-698 82006-700 82006-702 93000-040 93000-042 93000-044
VWR® Sample Library Tubes and Closures, 1.2 ml
Tubes Immunology Tubes
These 1.2 ml Library and Cluster Tubes are ideal for storage, dilution, mixing, harvesting, culture assays and screening based assays.
  • SBS footprint; 8×12 format to fit automated platforms
  • Available as individual tubes, strips of 8 tubes, and strips of 12 tubes
  • Tubes are made of virgin polypropylene and caps are made of polyethylene to provide a safe and secure seal
  • Certified free of detectable DNase, RNase, DNA, and PCR inhibitors
  • Sterile configurations are also pyrogren-free
Racks are printed with an alphanumeric grid, meet SBS standards, and can beMore Product Information