Axygen® Test Tubes, PS/PP, without Rim, Corning
Supplier: Corning
T-1275-PP T-1275-PS
89005-620CS 311.52 CAD
89005-620 89005-618
Axygen® Test Tubes, PS/PP, without Rim, Corning
Tubes Test Tubes
PS, clear or PP, transparent.
  • PP: good temperature resistance, good chemical resistance
  • PS: highly transparent, well suited to visual measurements
  • Matching push-in stoppers are optionally available
  • Can also be used as centrifuge tubes
Universal tubes for a wide range of laboratory and production applications.