Test Tubes without Rim
Supplier: United Scientific Supplies
TT9820-C TT9820-D TT9820-A TT9820-B TT9820-E TT9820-F TT9820-H TT9820-I TT9820-J TT9820-K TT9820-L
82024-104CS 832.55 CAD
82024-104 82024-106 76532-220 76532-222 76532-224 76532-226 76532-228 76532-230 76532-232 76532-234 76532-236
Test Tubes without Rim
Tubes Test Tubes
Pre-cleaned borosilicate glass test tubes without rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
  • Tubes do not have rims
  • Autoclavable
Rimless borosilicate glass test tubes are ideal for a variety of laboratory applications.
Order Now
SPECIFICATIONS
Environmentally Preferable