Kimble® Disposable Culture Tubes, Round Bottom, Screw-Cap Finish, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
73770-16100 73770-16150 73770-20150 73770-20125 73770-16125 73770-13100 73760-16125 90K16100F 73770-1690
60825-837CS 1032.63 CAD
60825-837 72690-020 72690-016 72690-018 72690-022 72690-024 72690-028 CA60826-153XFB 76212-366
Kimble® Disposable Culture Tubes, Round Bottom, Screw-Cap Finish, DWK Life Sciences
Tubes Culture Tubes
Tubes are designed for use in tissue culture work and general bacteriological applications.
  • Supplied without a marking area and without caps
  • Tubes are shrink-wrapped in plastic film in modular trays
  • Resistant to chemical, thermal and mechanical shock
  • Dishwasher safe and patented inner clamps keep tubes centered and vertical
Tubes are manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.
Please note: The "capacity" given below indicates the brim (overflow) capacity.