KIMAX® Test Tubes, Reusable, Borosilicate Glass, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
45042-1075 45042-13100 45042-15125 45042-16150 45042-18150 45042-20150 45042-25150 45042-25200
89000-840CS 1080.78 CAD
89000-840 89000-842 89000-844 89000-846 89000-848 89000-850 89001-430 89001-432
KIMAX® Test Tubes, Reusable, Borosilicate Glass, Kimble Chase
Tubes Culture Tubes
Test tubes are made from KG-33 glass tubing and feature a sturdy lip and uniform wall thickness for maximum heat transfer and chemical resistance.
Each tube has a durable white ceramic enamel marking spot. Tubes meet ASTM E982, Type I requirements.