KIMBLE® Test Tubes with Flat Head [ST] Stopper, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
898000-0005 898000-0010 898000-0025
KT898000-0005EA 88.58 CAD
KT898000-0005 KT898000-0010 KT898000-0025
KIMBLE® Test Tubes with Flat Head [ST] Stopper, DWK Life Sciences
Tubes Culture Tubes
Reusable test tubes with flat head stoppers.
  • Round bottom
  • Uniform wall thickness for maximum service
  • Solid, flat head standard taper stopper
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.