Gram's decolourizing solution
Supplier: G-Biosciences
Synonyms:
Gram's Decolorizer Solution, Gram's Decoloriser Solution
786-2085
786-2086
786-2087
77540-228EA
298.7
CAD
77540-228
77540-230
77540-232
Gram's decolourizing solution
Gram's decolorizing solution
Gram Decolorizer is a mixture of 25% Acetone and 75% Isopropanol solution.
- An aqueous solution of Acetone and Isopropanol
It is used for rapid decolorization before counterstaining