Nunc® Disposable Tubes, Round-Bottom, Polystyrene, Thermo Scientific
Supplier: Thermo Fisher Scientific
342919 343141 343923
CA62409-459CS 984.28 CAD
CA62409-459 CA62409-465 CA62409-467
Nunc® Disposable Tubes, Round-Bottom, Polystyrene, Thermo Scientific
Tubes Centrifuge Tubes
These transparent, polystyrene tubes offer a high degree of thermal and chemical stability.
Tubes withstand centrifuged forces up to 3000 ×g and are suitable for sample transport or the culture of microorganisms.
Ordering information: Additional polyethylene stoppers are sold separately.
Delivery information: Tube 62409-465 is sterilized and supplied with a mounted stopper.