Electrode Fill Solutions
Supplier: METTLER TOLEDO(INDUSTRIAL) CA
51343182 51343185 51343180 51343184 51343181
CA97001-718EA 46.01 CAD
CA97001-718 CA97001-710 CA97001-712 CA97001-714 CA97001-716
Electrode Fill Solutions
Electrolytes Electrode Filling Solutions
FRISCOLYT-B™ (84.4% glycerol, 11% KCl, and 4.6% H₂O) is for low temperature samples.
Delivery information: Supplied in 25 ml (0.9 oz.) bottles.