SP Wilmad-LabGlass Globe Style Separation Funnels with Glass Stopper and Stopcock, SP Industries
Supplier: SP Industries
LG-8420-112 LG-8420-106 LG-8420-108
14203-762EA 824.2 CAD
14203-762 14204-172 14206-096 14208-818 14209-018 14209-630 14210-064 14209-658 14210-316
SP Wilmad-LabGlass Globe Style Separation Funnels with Glass Stopper and Stopcock, SP Industries
Funnels Separating Funnels
These borosilicate glass pear-shaped separatory funnels are supplied with a Standard Taper glass stopper and a Standard Taper glass stopcock.