PYREX® Extraction Apparatus, with Friedrichs Condenser, [ST] Joints, Corning
Supplier: Corning
3880-M 3880-MCO
27614-105CS 1869.42 CAD
27614-105 27614-160
PYREX® Extraction Apparatus, with Friedrichs Condenser, [ST] Joints, Corning
Extractors Soxhlet and Soxhlet-Type Extractors
An efficient extraction apparatus for all general purposes
Consists of an extractor, a special Friedrichs-type condenser, and a flask, all equipped with [ST] joints. [ST] 24/40 between extraction and 250mL flask. [ST] 45/50 between extraction and condenser. Condenser tubulation O.D. is 10mm.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Monterrey