KIMBLE® RAY-SORB® Amber Separatory Funnels, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
29052F-250 29052F-125 29052F-500
30353-032EA 602.48 CAD
30353-032 30353-010 30353-054
KIMBLE® RAY-SORB® Amber Separatory Funnels, DWK Life Sciences
Funnels Separating Funnels
Designed with large lower stem I.D
for more complete separation. RAY-SORB® processed to protect materials sensitive to light of shorter wavelengths. Color-coded stopper. Designed from ASTM Specification E1096, Type IV requirements.