VWR® Premium Laboratory Refrigerators
Supplier: VWR International
77574-966 77574-968
77574-966EA 28508.13 CAD
77574-966 77574-968
VWR® Premium Laboratory Refrigerators
Refrigerators
Transform your laboratory environment with VWR® Premium laboratory refrigerators.
  • Ultra-quiet variable speed compressors
  • Microprocessor temperature control
  • Superior energy efficiency
  • Full array of alarms
  • Natural hydrocarbon refrigerants
These units bring the epitome of future-focused engineering to your scientific work.
Experience temperature control at its finest with precision microprocessor technology. Noisy disruptions can be avoided with ultra-quiet operation. Dynamic compressors reduces energy consumption by adapting to cooling demands.
Enhance your results and embrace a serene environment with VWR® Premium laboratory refrigerators. Step into a new era of tranquility and efficiency in your laboratory.
Certifications: ETL/cETL Listed.