LabKlenz™ 100 Alkaline Detergents
Supplier: Steris
1L1002 1L1005 1L1008 1L10D9
CA89089-192EA 1921.78 CAD
CA89089-192 CA89089-194 CA89089-196 77531-226
LabKlenz™ 100 Alkaline Detergents
Cleaning Agents Detergents and Surface Cleansers
LabKlenz™ 100 Detergent is a phosphate-free, liquid, alkaline detergent for use in automated laboratory glassware washing applications. It is formulated with corrosion inhibitors for enhanced materials compatibility.
  • Flexibility - Highly alkaline for effective removal of residues in both soft and hard water
  • Efficacy - Contains corrosion inhibitors for excellent materials compatibility
  • Cost savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - phosphate-free formulation with
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable