Hydrogen Peroxide WFI Sterile Solutions, 6%
Supplier: Steris
1S0577WR 1S0596WR 1S0508WR
CA75780-042CS 786.68 CAD
CA75780-042 CA75780-044 CA75780-046
Hydrogen Peroxide WFI Sterile Solutions, 6%
Cleaning Agents Detergents and Surface Cleansers
STERIS' 6% Hydrogen Peroxide WFI sterile solution removes residue from hard surfaces even from the most critical surfaces. It is formulated to be a low residue cleaner with minimal organic carbon.
  • Quality - Extremely low level of endotoxins from WFI produced by a Finn-Aqua® Multiple Effect Water Still
  • Ease of Use - Ready to use sterile hydrogen peroxide solution with extremely low odor and residue
  • Convenience - Double bagged for easy introduction into your aseptic areas
  • Safety - 0.2 µm filtered, aseptically filled and packaged minimizes potential for
More Product Information