ProKlenz® 120 High Performance Alkaline Detergent

Supplier: Steris
ProKlenz®
143205WR 143201WR
77531-586EA 516.19 CAD
77531-586 77531-598
ProKlenz® 120 High Performance Alkaline Detergent
Cleaning Agents Detergents and Surface Cleansers
CIP, COP, and manual cleaning.

  • pH (1%): 11.5, typical
  • Formulated alkaline detergent for effective removal of residues in both soft and hard water
  • Formulated for excellent materials compatibility
  • Reduces utility consumption and cleaning time with low foaming, free-rinsing formulation
  • Phosphate-free formulation with biodegradable surfactants
  • Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility

ProKlenz® 120 Alkaline detergent is a phosphate-free, liquid, alkaline detergent for use in clean-in-place (CIP), clean-out-of-place (COP), and manual cleaning applications. It is formulated for enhanced materials compatibility.

ProKlenz® 120 Alkaline detergent is specially formulated to deliver effective cleaning with excellent substrate compatibility in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

CAS: 64-02-8; 1310-58-3; 1310-73-2
UN 1814
Hazard class: 8
Packing group: II

Caution: Danger, GHS05; GHS07.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR